Cole-Parmer® Porcelain Desiccator Plates
Thermal shock and chemical resistant
  • Maximum temperature limit for use: 1000 °C
  • Resistance to acids and alkalis
The porcelain desiccator plates feature numerous 5 mm perforations along with a 25 mm center hole. Glazed on one side, the plates average coefficient of expansion from 20 to 200 °C is 3.56” x 10-5, gradually increasing to 4.69 x 10-5 at 1050 °C
5,900.00 - ₹6,900.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (InchShort) Material Description
Availability Pricing
COMPARE
7.5 Porcelain Porcelain Desiccator Plate, 190 mm dia
₹5,900.00INR / Each
COMPARE
9 Porcelain Porcelain Desiccator Plate, 230 mm dia
₹6,900.00INR / Each