Cole-Parmer® Porcelain Crucible Cover
Keep your samples protected
  • Resistant to acids and alkalies
  • Autoclavable
These replacement porcelain crucible covers are glazed with the exception of the rim. They can withstand temperatures up to 1150 °C (2102 °F). In addition, the covers are resistance to acids and alkalies except for hydrofluoric acid.
The correct size crucible cover is the next size larger than the diameter of your crucible. Compatible with the Cole-Parmer® porcelain wide-form, low-form, high-form, and Gooch filter crucibles.
7,600.00 - ₹14,400.00INR / Pkg of 12
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Autoclavable Cap Material Inside Rim Diameter (mm)
Availability Pricing
COMPARE
Yes Porcelain 26
₹14,400.00INR / Pkg of 12
COMPARE
Yes Porcelain 45
₹11,800.00INR / Pkg of 12
COMPARE
Yes Porcelain 54
₹7,600.00INR / Pkg of 6