Cole-Parmer® Essentials Buchner Funnels, Porcelain
Exceptional chemical resistance at an economical price
  • Thermal shock resistance
  • Autoclavable
Funnel includes fixed perforated plate and is glazed inside and out, except for the rim. Resistance to acids and alkalies is excellent, except for hydrofluoric acid.
1,000.00 - ₹40,700.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Filter Paper Diameter (mm) Overall Height (mm)
Availability Pricing
COMPARE
3 13 66
₹1,000.00INR / Each
COMPARE
35 40 91
₹1,800.00INR / Each
COMPARE
100 55 103
₹2,700.00INR / Each
COMPARE
200 70 140
₹3,300.00INR / Each
COMPARE
350 90 162
₹4,300.00INR / Each
COMPARE
600 110 194
₹5,800.00INR / Each
COMPARE
800 125 200
₹8,200.00INR / Each
COMPARE
1150 150 220
₹15,500.00INR / Each
COMPARE
2000 180 272
₹22,700.00INR / Each
COMPARE
4800 240 352
₹40,700.00INR / Each
COMPARE
3 13 66
₹21,200.00INR / Pkg of 12
COMPARE
35 40 91
₹20,300.00INR / Pkg of 12
COMPARE
100 55 103
₹33,000.00INR / Pkg of 8
COMPARE
200 70 140
₹18,500.00INR / Pkg of 6
Showing 1- 14 of 14