Cole-Parmer® Plastic Well Microscope Slides
Enables mounting of large specimens directly on slides
  • Reusable plastic slides protect against breakage
  • Deep well tapers from 2 mm on the edges to 3 mm in the center
These Cole-Parmer® Essentials well microscope slides are made of clear plastic and measure 3” x 1”. Each slide features one well that is approximately 16 mm in diameter with a depth of 2 mm on the edges to 3 mm in the center.
Choose from standard slides or large slides with a raise edge design for easy handling.
700.00 - ₹1,000.00INR / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Size Slide Type Description
Availability Pricing
COMPARE
3" x 1" 1 Well Microscope Slide with Well, Plastic, 3" x 1"; 10/Pk
₹1,000.00INR / Pkg of 10
COMPARE
3" x 1" with Raised Edges 1 Well Microscope Slide with Well, Plastic, 3" x 1" with Raised Edges; 10/Pk
₹700.00INR / Pkg of 10