Cole-Parmer® Piscean Infection Detection Kits for qPCR
Quickly and accurately detect the presence of piscean infections
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
161,000.00 - ₹206,000.00INR / Each
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Botrytis cinerea No DNA
₹161,000.00INR / Each
COMPARE
Botrytis cinerea Yes DNA
₹206,000.00INR / Each
COMPARE
Mycobacterium marinum and M. ulcerans No DNA
₹206,000.00INR / Each
COMPARE
Mycobacterium marinum and M. ulcerans Yes DNA
₹197,000.00INR / Each
COMPARE
Aleutian Disease Virus No DNA
₹161,000.00INR / Each
COMPARE
Aleutian Disease Virus Yes DNA
₹206,000.00INR / Each
COMPARE
Camelpox Virus No DNA
₹161,000.00INR / Each
COMPARE
Camelpox Virus Yes DNA
₹206,000.00INR / Each
COMPARE
Cyprinid Herpesvirus 3 No DNA
₹161,000.00INR / Each
COMPARE
Cyprinid Herpesvirus 3 Yes DNA
₹206,000.00INR / Each
COMPARE
Epizootic Hemorrhagic Disease Virus No RNA
₹161,000.00INR / Each
COMPARE
Epizootic Hemorrhagic Disease Virus Yes RNA
₹206,000.00INR / Each
COMPARE
Infectious Hematopoietic Necrosis Virus No RNA
₹161,000.00INR / Each
COMPARE
Infectious Hematopoietic Necrosis Virus Yes RNA
₹206,000.00INR / Each
COMPARE
Spring Viremia of Carp Virus No RNA
₹161,000.00INR / Each
COMPARE
Spring Viremia of Carp Virus Yes RNA
₹206,000.00INR / Each
COMPARE
Taï Forest ebolavirus No RNA
₹161,000.00INR / Each
COMPARE
Taï Forest ebolavirus Yes RNA
₹206,000.00INR / Each
COMPARE
Viral Hemorrhagic Septicemia Virus No DNA
₹161,000.00INR / Each
Showing 1- 19 of 19