Cole-Parmer® Pharmaprene® 3-Stop Pump Tubing
Exhibits very good chemical resistance to alcohols, saline solutions, and corrosive aqueous fluids
  • Stops ensure a perfect fit every time
  • Color-coded stops for easy identification
  • Tough but flexible for use with peristaltic pumps
  • Free of latex and animal-derived components and RoHS compliant
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Specially formulated Pharmaprene® tubing is flexible yet tough offering excellent pump life in peristaltic pumps. This durable tubing displays great chemical resistance to alcohols, corrosives, and saline solutions and exhibits good abrasion resistance and anti-aging properties under exposure to sunlight. It handles a wide operational temperature range from -50 to 135°C (-58 to 275°F) and shows excellent environmental stability even under changing temperatures to provide consistent and precise flow rates.
15,600.00 - ₹18,800.00INR / Pkg of 12
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
COMPARE
Orange/Green 0.38
₹15,700.00INR / Pkg of 12
COMPARE
Black/Black 0.76
₹18,600.00INR / Pkg of 12
COMPARE
Orange/Orange 0.89
₹15,600.00INR / Pkg of 12
COMPARE
White/White 1.02
₹18,800.00INR / Pkg of 12
COMPARE
Red/Red 1.14
₹15,700.00INR / Pkg of 12
COMPARE
Gray/Gray 1.3
₹15,600.00INR / Pkg of 12
COMPARE
Yellow/Blue 1.52
₹17,600.00INR / Pkg of 12
COMPARE
Blue/Blue 1.65
₹15,700.00INR / Pkg of 12
COMPARE
Green/Green 1.85
₹15,700.00INR / Pkg of 12
COMPARE
Purple/Purple 2.06
₹15,600.00INR / Pkg of 12
COMPARE
Purple/Black 2.29
₹15,600.00INR / Pkg of 12
COMPARE
Purple/White 2.79
₹15,700.00INR / Pkg of 12
Showing 1- 12 of 12