Cole-Parmer® Pestle, 99.5% Alumina
Get precise elemental analysis with minimal contamination
  • Chemical and heat resistant
  • Non absorbent surface
  • Autoclavable
Ideal for precise elemental analysis where minimal sample contamination is required. Pestles are glazed all over except for grinding surfaces. The alumina composition results in minimum sample contamination.
Mortar sold separately.
8,000.00 - ₹42,700.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Pestles Length (mm)
Availability Pricing
COMPARE
57
₹8,000.00INR / Each
COMPARE
57
₹42,700.00INR / Pkg of 3
COMPARE
100
₹22,200.00INR / Each
COMPARE
117
₹16,300.00INR / Each