Cole-Parmer® Open Ring Clamps
Provide support for your specific laboratory equipment
Ideal for supporting leveling bulbs and funnels safely, the open end design allows easy placement and removal. Choose from uncoated or PVC coated rings for added glassware protection.
NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.
6,000.00 - ₹37,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clamp Material Diameter (in) Length (in)
Availability Pricing
COMPARE
Aluminum 3 5.2
₹6,000.00INR / Each
COMPARE
PVC-coated Aluminum 3 12.91
₹25,200.00INR / Each
COMPARE
Aluminum 4 6.22
₹7,600.00INR / Each
COMPARE
PVC-coated Aluminum 4 15.91
₹28,700.00INR / Each
COMPARE
Aluminum 5 7.2
₹10,700.00INR / Each
COMPARE
PVC-coated Aluminum 5 16.81
₹37,800.00INR / Each