Cole-Parmer Nylon Chormatography Syringe Filters
Nylon66, is strong and heat resistant - ideal for HPLC and Chromatography solvents
  • Bidirectional flow with female luer lock on one side and a male luer slip on the other
  • Withstand pressure up to 100 psi (7 bar)
  • Batch test certificate included with each pack
Nylon Syringe Filters are most often used in HPLC and other analytical applications for either aqueous or organic mobile phase separation. Filters do not contain any adhesive, detergent, or surfactants so they have low extractables—great for research applications. Maximum use temperature is 50°C (122°F).
2,300.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Membrane Material Pore Size (µm)
Availability Pricing
COMPARE
13 Nylon 0.2
₹2,300.00INR / Each
COMPARE
13 Nylon 0.45
₹2,300.00INR / Each
COMPARE
25 Nylon 0.2
₹2,300.00INR / Each
COMPARE
25 Nylon 0.45
₹2,300.00INR / Each
COMPARE
33 Nylon 0.2
₹2,300.00INR / Each
COMPARE
33 Nylon 0.45
₹2,300.00INR / Each