Cole-Parmer® NPT(M) to Hose Barb Fittings, Straight Adapter, Natural Polycarbonate
Easily transition from threaded connection to soft tubing
  • Sharp back-barb securely holds tubing in place
  • Transparent fittiings—easily view fluid flow
  • Meets FDA 21 CFR 177.1580 and NSF 51 requirements, DEHP free, and RoHS compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Threaded barb adapters provide a quick way to transition from soft tubing to a system that accepts a threaded connection. Threads provide a tight and secure connection; a specially designed sharp back-barb expands tubing 40 to 50% above its normal ID for a leak-free, clamp-free seal up to 125 psi. Transparent fittings make it easy to verify fluid flow.
1,200.00 - ₹1,300.00INR / Pkg of 10
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
COMPARE
1/16 1/16
₹1,200.00INR / Pkg of 10
COMPARE
1/16 1/8
₹1,200.00INR / Pkg of 10
COMPARE
3/32 1/16
₹1,200.00INR / Pkg of 10
COMPARE
3/32 1/8
₹1,200.00INR / Pkg of 10
COMPARE
1/8 1/16
₹1,200.00INR / Pkg of 10
COMPARE
1/8 1/8
₹1,200.00INR / Pkg of 10
COMPARE
5/32 1/8
₹1,200.00INR / Pkg of 10
COMPARE
3/16 1/16
₹1,200.00INR / Pkg of 10
COMPARE
3/16 1/8
₹1,200.00INR / Pkg of 10
COMPARE
1/4 1/8
₹1,300.00INR / Pkg of 10
Showing 1- 10 of 10