Cole-Parmer® MSU-100 Compound Microscope
Robust and sturdy microscope that provides crisp, high-resolution images
  • Binocular 45° inclined tubes with 360° rotatable head
  • NeoLED illumination system increases light output
  • Coarse adjustment with friction control
  • Internal rechargeable batteries with an external battery charger and mains adapter for added convenience
The Cole-Parmer binocular microscope feature wide field eyepieces WF10x/18 mm with eyecups. The head features an adjustable interpupillary distance between 48 and 75 mm and a ±5 diopter adjustment on left tube. Featuring a 115 x 100mm mechanical stage with 55 x 22mm X-Y translation. Supplied with an external 100-240 V mains adapter/charger and rechargeable batteries for corded or cordless use.
104,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Head Configuration Magnification Power
Availability Pricing
COMPARE
Binocular 40x, 100x, 400x and 1000x Battery and VAC powered
₹104,000.00INR / Each