Cole-Parmer® MS Certified Vial Kits
Decrease your risk of failed analysis and sample reruns
  • Vial kits combine vials and caps with septa for your convenience
  • Constructed with silicone/PTFE septa for contaminant free sampling
  • Vials and caps are lot tested by HPLC for cleanliness
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Vial kits are available HPLC/GC certified or LC/MS and GC/MS certified. Each batch of HPLC/GC certified kits is tested on 15 critical parameters and each lot of LC/MS and GC/MS kits have been tested by LC/MS and GC/MS on traces of blank values and contaminations.
Short thread vials have a 9 mm polypropylene cap with silicone/PTFE septa and are constructed of clear or amber glass with a label and filling lines.
8,200.00 - ₹14,100.00INR / Case of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Septum Type Special Features Tube Color
Availability Pricing
COMPARE
Silicone White/PTFE Red HPLC/GC Certified Clear
₹8,200.00INR / Case of 100
COMPARE
Silicone Blue/PTFE Natural LC/MS and GC/MS Certified Clear
₹13,500.00INR / Case of 100
COMPARE
Silicone Blue/PTFE Natural LC/MS and GC/MS Certified Amber
₹14,100.00INR / Case of 100