Cole-Parmer® Mortar, 99.5% Alumina
Get precise elemental analysis with minimal contamination
  • Chemical and heat resistant
  • Non absorbent surface
  • Autoclavable
Ideal for precise elemental analysis where minimal sample contamination is required. The octagon-shaped alumina mortars are glazed all over except for grinding surfaces and the outside bottom. The alumina composition results in minimum sample contamination.
Pestle sold separately.
20,400.00 - ₹108,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Mortars Capacity (mL)
Availability Pricing
COMPARE
7
₹20,400.00INR / Each
COMPARE
7
₹108,000.00INR / Pkg of 3
COMPARE
80
₹54,100.00INR / Each