Cole-Parmer® Modular pH and ORP Electrodes
Universal sensor body, PPS construction. Models available for pH, ORP, low ionic strength solutions, and HF-resistant electrodes for solutions that should not exceed 1% hydrofluoric acid. Order pH electrode adapter at left to connect the new ERP-series electrodes to olderstyle cartridge electrode cap cable installations.
  • User-configurable self-cleaning, flat-surfaced electrodes for almost any application
  • Extended Reference Path (ERP™) technology for long life and stability
  • Modular plug-and-play design
  • Optional plug-in electronics produce amplified or mA signals
These cartridge-style electrodes are the next generation of the well-proven Cole-Parmer cartridge electrodes. The userconfigurable, plug-and-play nature of the system provides the ultimate in flexibility and versatility—purchase as few, or as many components needed for your application. If your application needs later change, single components can be added or subtracted, saving time and money of having to replace an entire system.
The reference system of the ERP-series has been enhanced with severe-duty reference gels to provide protection against thermal and chemical breakdown. In addition, a complex double-junction pathway protects the reference cell from contamination from heavy metals, sulfur, iodine, or proteins. Systems are configurable to interface with virtually any make or model
62,700.00 - ₹76,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Standard Application, Cartridge-Style, Double Junction, pH Electode
₹62,700.00INR / Each
COMPARE
HF-resistant, Cartridge-Style, Double Junction, pH Electode
₹70,900.00INR / Each
COMPARE
Standard Application, Cartridge-Style, Double Junction ORP Electode
₹76,300.00INR / Each