Cole-Parmer® Modular Magnetic Stirrer
Optimize your stirring needs and workspace with this completely customizable stirrer.
  • Attach up to 10 stir plates based on need
  • Simple touch and slide operation
  • LCD display with real time parameters
  • Corrosion-resistant glass stir top
  • Waterproof USB attachments
This unique multiposition stirrer allows you to decide how many stir plates to attach (up to 10) based on your mixing needs. Through direct USB connection or with the included cables it is simple to attach or remove a stir plate and configure to suit your work area. Each stir plate edge lights up to allow for accurate positioning of the vessel. Multiple stirring options allow a choice between one direction or reversing every 45 seconds for thorough mixing. Built in timer can be set for up to 9 hours or continuous operation.
19,400.00 - ₹99,400.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Power (VAC) Top Plate Diameter (in)
Availability Pricing
COMPARE
1500 100 to 240 3 5/8
₹99,400.00INR / Each
COMPARE
1500 100 to 240
₹21,800.00INR / Each
COMPARE
1500 100 to 240 3 19/32
₹19,400.00INR / Each