Cole-Parmer® Microscope Slides
Won’t stick or scratch
  • Easy to write on
  • Manufactured in accordance with ISO 8037/1
  • Measure 26 x 76 mm and 1 mm thick
  • Tolerance for thickness: ±0.05 mm
  • Planarity under 30 µm measured over 60 mm length
  • Choose from plain, frosted, or pink color frost options
Our slides feature precision ground edges on all four sides. Use plain microscope slides in general purpose applications. The raised coating on the Colorfrost slides keeps them separated to prevent sticking and scratching and provides a smooth writing surface for easy labeling. Color frosted slides make identification a snap. Manufactured from extra-white soda-line glass with very low iron content.
80,900.00INR / Case of 1440
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Slide Type Description
Availability Pricing
COMPARE
Plain Microscope Slides, Plain; 1440 slides
₹80,900.00INR / Case of 1440