Cole-Parmer® Micro Aperture Stoppers
Tapered to work on openings from 4 to 12.7 mm
  • Low sulfur content and low extractables
  • Simply cut to size
Use the tapered micro stoppers for openings from 4 to 12.7 mm. Available in soft, natural red rubber, or heat-resistant, green neoprene. Materials feature low sulfur content and low extractables. Cut to size.
27,600.00 - ₹33,900.00INR / Bag of 24
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material
Availability Pricing
COMPARE
Rubber
₹33,900.00INR / Bag of 24
COMPARE
Neoprene
₹27,600.00INR / Bag of 24