Cole-Parmer® Meat Speciation Detection Kits for qPCR
Quickly and accurately verify meat speciation in a food source
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Great value at 100 tests per kit
Use the speciation kits to clarify sources and types of products where accurate results are critical. Order a complete kit with mastermix or select a kit without mastermix and use your own.
161,000.00 - ₹230,000.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Anas platyrhynchos (Duck) No DNA
₹206,000.00INR / Each
COMPARE
Anas platyrhynchos (Duck) Yes DNA
₹230,000.00INR / Each
COMPARE
Capra hircus (Goat) No DNA
₹161,000.00INR / Each
COMPARE
Capra hircus (Goat) Yes DNA
₹206,000.00INR / Each
COMPARE
Equus caballus (Horse) No DNA
₹161,000.00INR / Each
COMPARE
Equus caballus (Horse) Yes DNA
₹206,000.00INR / Each
COMPARE
Gallus gallus (Chicken) No DNA
₹161,000.00INR / Each
COMPARE
Gallus gallus (Chicken) Yes DNA
₹206,000.00INR / Each
COMPARE
Meleagris gallopavo (Turkey) No DNA
₹161,000.00INR / Each
COMPARE
Meleagris gallopavo (Turkey) Yes DNA
₹209,000.00INR / Each
COMPARE
Ovis aries (Sheep) No DNA
₹161,000.00INR / Each
COMPARE
Ovis aries (Sheep) Yes DNA
₹206,000.00INR / Each
COMPARE
Sus scrofa (Pig/Pork) No DNA
₹161,000.00INR / Each
COMPARE
Universal Meat Detection No DNA
₹161,000.00INR / Each
COMPARE
Universal Meat Detection Yes DNA
₹206,000.00INR / Each
Showing 1- 15 of 15