Cole-Parmer® LB-400 Series Analytical Balances
Quality and precision at an economical price
  • Five-year industry-leading warranty
  • = 3 seconds stabilization time
  • 0.01mg and 0.1mg four-place readability
  • Easily adjustable filter settings
  • Internal and external calibration options
  • Large, backlit LCD for improved visibility of weighing results
  • USB, RS-232, and Wi-Fi† interfaces
  • Multiple working modes handle a variety of weighing functions
  • Spacious weighing chamber allows for tool-free disassembly and assembly
  • Anti-static coating on weighing chamber panes
The LB-400 analytical balances provide accurate measurements of weighed loads up to a maximum of 62g to 320g. A spacious three-door glass draft shield ensures weighing is as precise as possible, protecting the balance from external factors such as dust and drafts that could affect measurement accuracy. Internal and external calibration options allow you to choose the calibration method that best suits your specific needs, ensuring the balances remain accurate over time.
The large backlit LCD display provides clear and easy-to-read weighing results which is helpful in sub-optimal lighting conditions. The balances come equipped with a filter level setting that is easily adjusted to either a fast or slow mode, allowing you to tailor the balance's response to the working environment.
Quick access keys, balance taring, balance zeroing, auto switch-off, and backlight turn-off time are all incorporated into the design to enhance user-friendliness. Record important information such as users, products, weighings, and tares to simplify data management and retrieval, aiding in keeping track of measurement history and relevant details.
†Only models with internal calibration feature Wi-Fi interface.
422,000.00 - ₹876,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Description
Availability Pricing
COMPARE
Internal 220 Analytical Balance with LCD, 82g/220g x 0.01mg/0.1mg, Internal Calibration
₹876,000.00INR / Each
COMPARE
Internal 160 Analytical Balance with LCD, 160g x 0.1mg, Internal Calibration
₹543,000.00INR / Each
COMPARE
Internal 220 Analytical Balance with LCD, 220g x 0.1mg, Internal Calibration
₹515,000.00INR / Each
COMPARE
Internal 310 Analytical Balance with LCD, 310g x 0.1mg, Internal Calibration
₹601,000.00INR / Each
COMPARE
External 62 Analytical Balance with LCD, 62g x 0.1mg, External Calibration
₹425,000.00INR / Each
COMPARE
External 160 Analytical Balance with LCD, 160g x 0.1mg, External Calibration
₹422,000.00INR / Each
COMPARE
External 220 Analytical Balance with LCD, 220g x 0.1mg, External Calibration; 100/240 VAC
₹425,000.00INR / Each
COMPARE
External 310 Analytical Balance with LCD, 310g x 0.1mg, External Calibration
₹456,000.00INR / Each
Showing 1- 8 of 8