Cole-Parmer® Industrial Silicone-Filled Bimetal Thermometers
Improve readings and response times with silicone-filled dial
  • Silicone-filled for reduction of pointer flutter on high vibration applications
  • Easy-to-read dual °F/°C scales provide enhanced flexibility
  • Anti-parallax dished dial for superior readability
  • Durable all-welded stainless steel construction for tough environments
  • Prevent dial fog, entry of moisture, interior corrosion, and coil freeze up with hermetically sealed case
  • Rugged NEMA 4X/IP66 protection
  • Recalibrate using the external reset screw
  • ±1% full-scale accuracy according to ASME B40.200 (B40.3 Grade A)
The Cole-Parmer® silicone-filled industrial bimetal thermometers feature NEMA 4X/IP66 protection for tough applications. Designed for the most severe vibration conditions, the silicone is used to dampen and lubricate internal parts, enhancing readability and lifespan. Silicone-filled thermometers offer superior performance that will hold up to the toughest vibration applications. The lens material is constructed of polycarbonate due to the expansion properties of silicone.
Accuracy is ±1% full-scale when stem is immersed 2" in liquids or 4" in gases (immerse models with a 2.5" stem length 2" in gases). Stem length is measured from the top of the threads to the bottom of the stem tip. Stem diameter is 0.25” with a ½” NPT(M) back connection. Maximum over temperature for thermometers with a max temperature of 250°F is 100% of span, and thermometers with a max temperature of 550°F is 50% of span. The ambient temperature range limits are –40°F to 150°F (–40°C to 65°C).
19,400.00 - ₹20,600.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Dial Size (in) Max Temperature (° F) Stem Length (in)
Availability Pricing
COMPARE
3 250 2.5
₹19,400.00INR / Each
COMPARE
3 250 4
₹19,400.00INR / Each
COMPARE
3 250 6
₹19,400.00INR / Each
COMPARE
3 250 9
₹19,400.00INR / Each
COMPARE
3 250 12
₹19,400.00INR / Each
COMPARE
3 550 4
₹19,400.00INR / Each
COMPARE
3 550 6
₹19,400.00INR / Each
COMPARE
5 250 2.5
₹20,600.00INR / Each
COMPARE
5 250 6
₹20,600.00INR / Each
Showing 1- 9 of 9