Cole-Parmer® Industrial Conductivity Cells
Rugged sensors withstand high pressure and temperature
  • Designed for in-line or submersion conductivity measurements in processes that generate up to 250 psi and 392°F (200°C)
  • 316 stainless steel construction provides excellent chemical compatibility
  • Available with automatic temperature compensation (ATC) for compatibility with most makes and models of conductivity controllers
Cole-Parmer industrial conductivity cells are designed for high performance in harsh measurement conditions. Stainless steel construction resists aggressive chemicals, high pressures, and high temperatures. Threaded ¾” NPT electrode bodies allow for in-line or submersion mounting. Cells are well-suited for boiler or ultrapure water applications.
107,000.00 - ₹108,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
High Temperature Conductivity Cell, K = 0.1, 100 Ohms Pt RTD ATC
₹108,000.00INR / Each
COMPARE
High Temperature Conductivity Cell, K = 1.0, 100 Ohms Pt RTD ATC
₹108,000.00INR / Each
COMPARE
High-Temp Extended-Length Conductivity Cell, K=1.0, RTD
₹108,000.00INR / Each
COMPARE
High Temperature Conductivity Cell, K = 10.0, No ATC
₹107,000.00INR / Each
COMPARE
High Temperature Conductivity Cell, K = 10, 100 Ohms Pt RTD ATC
₹108,000.00INR / Each