Cole-Parmer® Human Herpes Viral Infection Detection Kits for qPCR
Quickly and accurately detect the presence of human herpes virus
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
144,000.00 - ₹206,000.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Cytomegalovirus (HHV5) No DNA
₹161,000.00INR / Each
COMPARE
Cytomegalovirus (HHV5) Yes DNA
₹206,000.00INR / Each
COMPARE
Epstein Barr Virus (HHV4) No DNA
₹161,000.00INR / Each
COMPARE
Epstein Barr Virus (HHV4) Yes DNA
₹206,000.00INR / Each
COMPARE
Human Herpesvirus 3 Yes DNA
₹206,000.00INR / Each
COMPARE
Human Herpesvirus 6 No DNA
₹161,000.00INR / Each
COMPARE
Human Herpesvirus 6 Yes DNA
₹206,000.00INR / Each
COMPARE
Human Herpesvirus 7 No DNA
₹161,000.00INR / Each
COMPARE
Human Herpesvirus 8 No DNA
₹144,000.00INR / Each
COMPARE
Human Herpesvirus 8 Yes DNA
₹206,000.00INR / Each
Showing 1- 10 of 10