Cole-Parmer® Human Hepatitis Infection Detection Kits for qPCR
Quickly and accurately detect the presence of human hepatitis infection
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
161,000.00 - ₹206,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Hepatitis Delta Virus No RNA
₹161,000.00INR / Each
COMPARE
Hepatitis Delta Virus Yes RNA
₹206,000.00INR / Each
COMPARE
Hepatitis E Virus No RNA
₹161,000.00INR / Each
COMPARE
Hepatitis E Virus Yes RNA
₹206,000.00INR / Each