Cole-Parmer® Human Gastrointestinal Infection Detection Kits for qPCR
Quickly and accurately detect the presence of human gastrointestinal pathogens
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
155,000.00 - ₹328,000.00INR / Each
33 variations of this product are available.
See All Product Options Below
33 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Bacteroides (All Species) No DNA
₹206,000.00INR / Each
COMPARE
Bacteroides (All Species) Yes DNA
₹183,000.00INR / Each
COMPARE
Entamoeba (All Species) No DNA
₹161,000.00INR / Each
COMPARE
Entamoeba (All Species) Yes DNA
₹206,000.00INR / Each
COMPARE
Clostridium difficile (toxin B) No DNA
₹206,000.00INR / Each
COMPARE
Clostridium difficile (toxin B) Yes DNA
₹211,000.00INR / Each
COMPARE
Enteropathogenic Escherichia coli No DNA
₹161,000.00INR / Each
COMPARE
Enteropathogenic Escherichia coli Yes DNA
₹206,000.00INR / Each
COMPARE
Escherichia coli O104:H4 No DNA
₹161,000.00INR / Each
COMPARE
Escherichia coli O104:H4 Yes DNA
₹328,000.00INR / Each
COMPARE
Helicobacter pylori No DNA
₹161,000.00INR / Each
COMPARE
Helicobacter pylori Yes DNA
₹206,000.00INR / Each
COMPARE
Enterococcus faecalis No DNA
₹161,000.00INR / Each
COMPARE
Enterococcus faecalis Yes DNA
₹206,000.00INR / Each
COMPARE
Bifidobacterium bifidum No DNA
₹161,000.00INR / Each
COMPARE
Bifidobacterium bifidum Yes DNA
₹206,000.00INR / Each
COMPARE
Salmonella (All pathogenic species) No DNA
₹206,000.00INR / Each
COMPARE
Salmonella (All pathogenic species) Yes DNA
₹217,000.00INR / Each
COMPARE
Vibrio cholerae (All subspecies) No DNA
₹206,000.00INR / Each
COMPARE
Vibrio cholerae (All subspecies) Yes DNA
₹217,000.00INR / Each
Showing 1 - 20 of 33