Cole-Parmer® Hose Barb Fittings, Tee Union, Natural Polycarbonate
Easily combine or split fluid flow using tee fittings
  • Sharp back-barb securely holds tubing in place
  • Transparent fittiings—easily view fluid flow
  • Meets FDA 21 CFR 177.1580 and NSF 51 requirements, DEHP free, and RoHS compliant
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Whether you need to split a single fluid stream into two, combine two streams into one, or create a bypass, these tee unions offer the flexibility required for complex fluid routing. A specially designed sharp back-barb expands tubing 40 to 50% above its normal ID for a leak-free, clamp-free seal up to 125 psi. Transparent fittings make it easy to verify fluid flow.
1,500.00 - ₹1,600.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
COMPARE
1/16 1/16 1/16
₹1,500.00INR / Pkg of 10
COMPARE
3/32 3/32 3/32
₹1,500.00INR / Pkg of 10
COMPARE
1/8 1/8 1/8
₹1,500.00INR / Pkg of 10
COMPARE
5/32 5/32 5/32
₹1,500.00INR / Pkg of 10
COMPARE
3/16 3/16 3/16
₹1,500.00INR / Pkg of 10
COMPARE
1/4 1/4 1/4
₹1,600.00INR / Pkg of 10