Cole-Parmer® Hose Barb Fitting, Y Union, HDPE
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These high-density polyethylene (HDPE) fittings are softer than nylon, polypropylene, and Kynar®. They exhibit excellent resistance to chemicals but have lower thermal resistance than other materials.
2,400.00 - ₹6,600.00INR / Pkg of 10
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
COMPARE
1/16 1/16 1/16
₹2,400.00INR / Pkg of 10
COMPARE
3/32 3/32 3/32
₹2,400.00INR / Pkg of 10
COMPARE
1/8 1/8 1/8
₹2,400.00INR / Pkg of 10
COMPARE
5/32 5/32 5/32
₹3,400.00INR / Pkg of 10
COMPARE
1/4 1/4 1/4
₹3,500.00INR / Pkg of 10
COMPARE
5/16 5/16 5/16
₹3,700.00INR / Pkg of 10
COMPARE
3/8 3/8 3/8
₹3,700.00INR / Pkg of 10
COMPARE
5/8 5/8 5/8
₹5,100.00INR / Pkg of 10
COMPARE
3/4 3/4 3/4
₹5,400.00INR / Pkg of 10
COMPARE
1 1 1
₹6,600.00INR / Pkg of 10
Showing 1- 10 of 10