Cole-Parmer® Hose Barb Fitting, Straight Union, HDPE
Engineered for secure, leak-resistant hose-to-hose connections
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • Meets FDA requirements 21 CFR 177.1520 (when used unmodified for the manufacture of food contact articles) and RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Easily connect two tubing with straight unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. These high-density polyethylene (HDPE) fittings are softer than nylon, polypropylene, and Kynar®. They exhibit excellent resistance to chemicals but have lower thermal resistance than other materials.
1,100.00 - ₹2,400.00INR / Pkg of 10
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in)
Availability Pricing
COMPARE
1/8 1/8
₹1,300.00INR / Pkg of 10
COMPARE
3/32 3/32
₹1,100.00INR / Pkg of 10
COMPARE
1/8 1/8
₹1,100.00INR / Pkg of 10
COMPARE
5/32 5/32
₹1,500.00INR / Pkg of 10
COMPARE
3/16 3/16
₹1,600.00INR / Pkg of 10
COMPARE
1/4 1/4
₹1,300.00INR / Pkg of 10
COMPARE
5/16 5/16
₹1,500.00INR / Pkg of 10
COMPARE
1/2 1/2
₹1,500.00INR / Pkg of 10
COMPARE
5/8 5/8
₹1,500.00INR / Pkg of 10
COMPARE
3/4 3/4
₹1,900.00INR / Pkg of 10
COMPARE
1 1
₹2,400.00INR / Pkg of 10
Showing 1- 11 of 11