Cole-Parmer® Hose Barb Fitting, 90° Elbow, HDPE
Easily change the direction of the fluid flow with 90° elbows
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Easily connect two tubing with 90° elbow. A single barb on each end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on each port prevents tubing from rotating, minimizing kinks. These high-density polyethylene (HDPE) fittings are softer than nylon, polypropylene, and Kynar®. They exhibit excellent resistance to chemicals but have lower thermal resistance than other materials.
1,600.00 - ₹2,700.00INR / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in)
Availability Pricing
COMPARE
1/16 1/16
₹1,600.00INR / Pkg of 10
COMPARE
3/32 3/32
₹1,600.00INR / Pkg of 10
COMPARE
1/8 1/8
₹1,600.00INR / Pkg of 10
COMPARE
5/32 5/32
₹1,700.00INR / Pkg of 10
COMPARE
3/16 3/16
₹1,700.00INR / Pkg of 10
COMPARE
1/4 1/4
₹1,700.00INR / Pkg of 10
COMPARE
5/16 5/16
₹1,800.00INR / Pkg of 10
COMPARE
3/8 3/8
₹1,800.00INR / Pkg of 10
COMPARE
1/2 1/2
₹1,800.00INR / Pkg of 10
COMPARE
5/8 5/8
₹1,900.00INR / Pkg of 10
COMPARE
3/4 3/4
₹2,100.00INR / Pkg of 10
COMPARE
1 1
₹2,700.00INR / Pkg of 10
Showing 1- 12 of 12