Cole-Parmer® Horizontal Mounting Bars with Coupler
Connect rods and / or clamps to your frame or build a custom lab frame
  • Use with 1/2" (12.7mm) diameter frame rods
  • Threaded end for direct installation into channels. Comes with spring couple for use in steel 41mm (1.61") channels. Locking nut included. Use with steel channel frames.
This special connector fastens lab-frame rod to steel channels, by using a threaded end for direct installation into channel. Includes spring coupler and locking nut. (For use with steel channel 1 1/2" (41mm) in thickness)
8,600.00 - ₹9,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accepts Rod Diameter (in) Connector Material Length (in)
Availability Pricing
COMPARE
0.5 Nickel-plated Zinc 6
₹9,300.00INR / Each
COMPARE
0.5 Nickel-plated Zinc 8
₹8,600.00INR / Each