Cole-Parmer® HM-200 Series Heating Mantles
Thermal insulating cartridge provides maximum heat transfer
  • Chemical resistant polypropylene housing is "safe to the touch"
Quickly and safely heat round-bottom flasks to 450°C (842°F). These simple-to-use heating mantles feature a built-in power controller with dial markings from 1 to 10 for repeatability. An amber-colored "heater on" light and a clear-colored "power on" light indicate when the heating mantle is in use. The replaceable coiled heating element suspended within a thermal insulating cartridge provides maximum heat transfer and support.
Unique ventilation slots and ceramic fiber insulation keep the chemical-resistant polypropylene exterior cool even while in use. A safety earth (ground) screen and double fuses provide added safety.
87,400.00 - ₹192,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Max temperature (° F) Power (VAC)
Availability Pricing
COMPARE
50 842 230
₹87,400.00INR / Each
COMPARE
100 842 230
₹93,100.00INR / Each
COMPARE
250 842 230
₹97,200.00INR / Each
COMPARE
500 842 230
₹112,000.00INR / Each
COMPARE
1000 842 230
₹113,000.00INR / Each
COMPARE
2000 842 230
₹187,000.00INR / Each
COMPARE
3000 842 230
₹192,000.00INR / Each
COMPARE
5000 842 230
₹173,000.00INR / Each
Showing 1- 8 of 8