Cole-Parmer® High-Capacity Tube Racks
Excellent for high-throughput labs requiring large sample storage
  • Large capacity—eliminate the need to have multiple tube racks
  • Lightweight yet durable polypropylene construction
  • Alphanumeric identification provides quick sample location and retrieval
Easily store and transport samples in these sturdy, high-capacity tube racks. High capacity allows you to place various testing groupings into one rack and eliminate the need to purchase multiple racks.
14,400.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Max Tube Size (mm) Tubes Held
Availability Pricing
COMPARE
White 20 120
₹14,400.00INR / Each
COMPARE
Blue 20 120
₹14,400.00INR / Each
COMPARE
Blue 13 216
₹14,400.00INR / Each
COMPARE
White 13 216
₹14,400.00INR / Each
COMPARE
Blue 16 120
₹14,400.00INR / Each
COMPARE
White 16 120
₹14,400.00INR / Each