Cole-Parmer® High-Accuracy Compound Transmitters
Withstand high shock and vibration applications
  • Welded stainless steel (SS) housing rated to NEMA 4 (IP65) standards
  • Choose between 4 to 20 mA and 0.1 to 5.1 V output signal
Transducers feature SS capacitive sensor for your more demanding systems—ElectroMagnetic Interference (EMI)/Radio Frequency Interference (RFI) resistance ensures accurate and consistent performance in any environment. Adjustable zero and span potentiometers improve accuracy.
107,000.00 - ₹114,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Output
Availability Pricing
COMPARE
±0.13% full-scale 15 4 to 20 mA
Call For Price
COMPARE
±0.13% full-scale 60 4 to 20 mA
₹107,000.00INR / Each
COMPARE
±0.13% full-scale 100 4 to 20 mA
₹107,000.00INR / Each
COMPARE
±0.13% full-scale 15 0.1 to 5.1 VDC
₹114,000.00INR / Each
COMPARE
±0.13% full-scale 30 0.1 to 5.1 VDC
₹107,000.00INR / Each