Cole-Parmer® Heavy-Duty Laboratory Support Bases
  • Bases and rods are sold separately
Economical heavy-duty support base are made with a cast iron base and the rods made of either 303 stainless steel or aluminum. Stand features a corrosion-resistant, electro polished cast iron base. All bases feature a built-in support rod holder with locking knob that accepts 0.51” (1.3 cm) diameter support rod. Support bases and rods are each sold separately.
12,800.00 - ₹49,900.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Accepts Rod Diameter (in) Length (in) Width (in)
Availability Pricing
COMPARE
0.5 6 4
₹16,100.00INR / Each
COMPARE
0.5 8 5
₹26,100.00INR / Each
COMPARE
0.5 9 6
₹34,100.00INR / Each
COMPARE
0.5 10 8
₹49,900.00INR / Each
COMPARE
0.5 6 6
₹31,800.00INR / Each
COMPARE
0.5 7 7
₹12,800.00INR / Each