Cole-Parmer® Glove Boxes
Monitor humidity and protect materials – affordably!
  • Slanted window eliminates optical distortion
  • Spatially efficient with ample working chamber
  • Glove ports: 6" (15.2 cm) dia O-rings
  • Glove size: 6" (15.2 cm) dia cuffs x 24" (61 cm) L
Use these economical glove boxes on your crowded lab bench or in large walk-in areas. The wide front access door makes loading equipment easy. Clear 1⁄4"-thick acrylic access and pass box doors ensure good visibility. Painted steel construction and EPDM foam seals endures everyday use. The included chamber hygrometer monitors humidity from 20 to 95% RH. Two ball valves are for purging gas.
616,000.00 - ₹814,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Chamber Depth (in) Chamber Height (in) Chamber Width (in)
Availability Pricing
COMPARE
27 20 20
₹616,000.00INR / Each
COMPARE
32 24 20
₹814,000.00INR / Each