Cole-Parmer® Fluorimetry Cuvettes
For use with Jenway fluorimeters
34,400.00 - ₹257,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
100 µl Quartz Cell (3 Windows)
₹201,000.00INR / Each
COMPARE
500 µl Glass Cell (Minimum 320 nm, 4 Windows With PTFE Lid) (360 to 2500 nm)
₹108,000.00INR / Each
COMPARE
500 µl Quartz Cell (Minimum 200 nm, 4 Windows With PTFE Lid) (200 to 2500 nm)
₹185,000.00INR / Each
COMPARE
Macro Plastic 3 mL Cuvettes, 10 mm, Visible, 4 Windows, Pack of 10
₹41,700.00INR / Pkg of 100
COMPARE
10 x 10 mm Macro Glass Cell (UV, 4 Windows)
₹88,700.00INR / Each
COMPARE
10 x 10 mm Macro Glass Cell (Visible, 4 Windows)
₹34,400.00INR / Each
COMPARE
Semi-Micro Flow Cell (UV, 3 Windows)
₹257,000.00INR / Each
Showing 1- 7 of 7