Cole-Parmer® Fish Speciation Detection Kits for qPCR
Quickly and accurately verify the fish speciation in a food source
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Great value at 100 tests per kit
Use the speciation kits to clarify sources and types of products where accurate results are critical. Order a complete kit with mastermix or select a kit without mastermix and use your own.
161,000.00 - ₹206,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Universal Fish Detection No DNA
₹161,000.00INR / Each
COMPARE
Universal Fish Detection Yes DNA
₹206,000.00INR / Each