Cole-Parmer® Feline Infection Detection Kits for qPCR
Quickly and accurately detect the presence of feline infections
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
155,000.00 - ₹206,000.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Bartonella henselae No DNA
₹161,000.00INR / Each
COMPARE
Bartonella henselae Yes DNA
₹206,000.00INR / Each
COMPARE
Bordetella bronchiseptica and B. parapertussis No DNA
₹206,000.00INR / Each
COMPARE
Bordetella bronchiseptica and B. parapertussis Yes DNA
₹155,000.00INR / Each
COMPARE
Chlamydophila felis No DNA
₹161,000.00INR / Each
COMPARE
Chlamydophila felis Yes DNA
₹206,000.00INR / Each
COMPARE
Mycoplasma felis No DNA
₹161,000.00INR / Each
COMPARE
Mycoplasma haemofelis No DNA
₹161,000.00INR / Each
COMPARE
Mycoplasma haemofelis Yes DNA
₹206,000.00INR / Each
COMPARE
Toxoplasma gondii No DNA
₹161,000.00INR / Each
COMPARE
Feline Coronavirus No RNA
₹161,000.00INR / Each
COMPARE
Feline Herpesvirus No DNA
₹161,000.00INR / Each
COMPARE
Feline Herpesvirus Yes DNA
₹206,000.00INR / Each
COMPARE
Feline Immunodeficiency Virus No RNA
₹161,000.00INR / Each
COMPARE
Feline Immunodeficiency Virus Yes RNA
₹206,000.00INR / Each
Showing 1- 15 of 15