Cole-Parmer® FDA-Compliant Carboy w/ Spigot and Handles, PP
Get a better grip with two-handle construction
  • Spigot quickly dispenses solutions
  • Leakproof PP cap
  • Easy to read, molded graduations
  • Autoclavable at 250°F (121°C)
These rugged, durable plastic carboys allow you to meet your large volume liquid storage and dispensing needs. Integrated wide handles make them easy to carry and pour, even with gloves on. The attached spigot allows for simple and accurate dispensing of solutions. Round design maximizes mixing efficiency. Polypropylene (PP) cap with integrated seal ring ensures leakproof seal. Carboys are autoclavable at 250 °F (121 °C) and are chemical resistant.
22,700.00 - ₹47,300.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
COMPARE
10 83 mm Polypropylene(PP)
₹22,700.00INR / Each
COMPARE
20 83 mm Polypropylene(PP)
₹27,200.00INR / Each
COMPARE
50 83 mm Polypropylene(PP)
₹47,300.00INR / Each