Cole-Parmer® Extra-Large Laboratory Frame Sets
Corrosion resistant frames and rods
  • Custom fit any laboratory with the right frame kit for your application
Frame sets come prepackaged with rods, connectors, and lab frame feet to accommodate all your clamp and apparatus needs. For custom frame sets, choose items separately.
Extra Large Lab-Frame
Horizontal or vertical mounting. Ideal for complex glassware set-ups, base can be permanently mounted to a wall or floor for added stability
Frame Dimensions (W x H): 48 x 72” (121. x 182.9 cm)
Base leg dimension (D): 12” (30.5 cm)
412,000.00 - ₹603,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accepts Rod Diameter (in) Clamp Type Length (in)
Availability Pricing
COMPARE
₹412,000.00INR / Each
COMPARE
₹603,000.00INR / Each