Cole-Parmer® Essentials Vertical/Chest Aluminum Racks for Standard 2" Boxes
Aluminum racks offer better temperature uniformity in vapor phase in liquid nitrogen tanks
  • Lightweight aluminum construction for easier handling
  • Accommodates standard 2" cardboard, plastic, and metal boxes
  • Locking rod keeps sample storage boxes securely in the rack
Use these vertical racks with a wide variety of chest freezers and liquid nitrogen tanks. Dual handles make it easy to place racks in and out of freezers and nitrogen tanks.
16,500.00 - ₹27,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in)
Availability Pricing
COMPARE
9 boxes 19 13/16
₹21,900.00INR / Each
COMPARE
10 boxes 22
₹16,500.00INR / Each
COMPARE
11 boxes 24 3/4
₹17,300.00INR / Each
COMPARE
13 boxes 28 5/8
₹18,700.00INR / Each
COMPARE
14 boxes 30 3/4
₹27,000.00INR / Each
COMPARE
15 boxes 33
₹20,800.00INR / Each