Cole-Parmer® Essentials Sterile Erlenmeyer Flasks with Cap, Polycarbonate
Sterile Erlenmeyer flasks for mixing, storage, and other liquid handling processes
  • Gamma sterilized (SAL 10-6)
  • DNase/RNase free and nonpyrogenic
  • Withstands temperature range of -80 to 121 °C (-112 to 249.8 °F)
  • Liquid sticking-resistant neck for easier pouring
  • Includes HDPE cap; choice of plug seal or vented
Ideal for media preparation, mixing cell cultures, and storage, these sterile polycarbonate (PC) Erlenmeyer flasks are a safe and shatterproof alternative to glass. The clear flasks feature molded-in graduations allowing you quick volume assessment. Liquid sticking-resistant design at the neck repels liquid for easier pouring. These Erlenmeyer flasks have a temperature range of -80 to 121 °C (-112 to 249.8 °F), and the flask body can be autoclaved a single time. Optional baffled bottom models that help improve mixing are available.
HDPE caps are included. Choose from a leakproof plug seal cap or a vented cap with 0.22 µm hydrophobic PTFE membrane for continuous gas exchange.
Manufactured in an ISO Class 8 (formerly Class 100,000) cleanroom environment, these Individually packaged Erlenmeyer flasks are sterilized by gamma irradiation per ISO 11137-2 (SAL 10-6), and are ISO 13485 and ISO 9001 certified, DNase/RNase free, and nonpyrogenic.
10,200.00 - ₹26,700.00INR / Case of 24
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap Type Graduation Interval (mL)
Availability Pricing
COMPARE
125 Seal 25
₹15,000.00INR / Case of 24
COMPARE
125 Vent 25
₹16,400.00INR / Case of 24
COMPARE
250 Seal 25
₹10,200.00INR / Case of 12
COMPARE
250 Vent 25
₹11,400.00INR / Case of 12
COMPARE
500 Seal 50
₹13,500.00INR / Case of 12
COMPARE
500 Vent 50
₹15,300.00INR / Case of 12
COMPARE
1000 Seal 50
₹11,300.00INR / Case of 6
COMPARE
1000 Vent 50
₹12,000.00INR / Case of 6
COMPARE
2000 Seal 100
₹22,800.00INR / Case of 6
COMPARE
2000 Vent 100
₹23,400.00INR / Case of 6
COMPARE
3000 Seal 100
₹26,000.00INR / Case of 4
COMPARE
3000 Vent 100
₹26,700.00INR / Case of 4
COMPARE
125 Vent 25
₹21,700.00INR / Case of 24
COMPARE
250 Vent 25
₹15,300.00INR / Case of 12
COMPARE
500 Vent 50
₹20,000.00INR / Case of 12
COMPARE
1000 Vent 50
₹13,200.00INR / Case of 6
Showing 1- 16 of 16