Cole-Parmer® Equine Infection Detection Kits for qPCR
Quickly and accurately detect the presence of equine infections
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
Each kit is designed to ensure the broadest possible detection profile and detection of all relevant strains and subtypes. Order a complete kit with mastermix or a kit without mastermix and use your own mastermix.
161,000.00 - ₹206,000.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Binomial Name Mastermix Included Test Type
Availability Pricing
COMPARE
Babesia caballi Yes DNA
₹206,000.00INR / Each
COMPARE
Theileria equi No DNA
₹161,000.00INR / Each
COMPARE
Theileria equi Yes DNA
₹206,000.00INR / Each
COMPARE
African Horse Sickness Virus No RNA
₹161,000.00INR / Each
COMPARE
African Horse Sickness Virus Yes RNA
₹206,000.00INR / Each
COMPARE
Equid Herpesvirus 1 No DNA
₹161,000.00INR / Each
COMPARE
Equid Herpesvirus 1 Yes DNA
₹206,000.00INR / Each
COMPARE
Equid Herpesvirus 3 No DNA
₹161,000.00INR / Each
COMPARE
Equid Herpesvirus 3 Yes DNA
₹206,000.00INR / Each
COMPARE
Equid Herpesvirus 4 No DNA
₹161,000.00INR / Each
COMPARE
Equid Herpesvirus 4 Yes DNA
₹206,000.00INR / Each
COMPARE
Equine Infectious Anemia Virus No RNA
₹161,000.00INR / Each
COMPARE
Equine Infectious Anemia Virus Yes RNA
₹206,000.00INR / Each
COMPARE
Equine/Canine Influenza (H3N8 & H7N7) No RNA
₹161,000.00INR / Each
COMPARE
Equine/Canine Influenza (H3N8 & H7N7) Yes RNA
₹206,000.00INR / Each
Showing 1- 15 of 15