Cole-Parmer® Enlarging Check Valves, Polypropylene
Safeguard your fluid system from backflows while transitioning diameters
  • Polypropylene body with silicone diaphragm offers broad chemical compatibility
  • Free-floating, disk diaphragm allows installation in any position
  • 0.5 psi breaking pressure
  • 1.9 psi sealing pressure
  • Works with both liquids and gases
  • USP Class VI, FDA 21 CFR 177.1520 compliant for biocompatibility
  • Animal-derivative, DEHP, and BPA free and RoHS compliant
Place these check valves in your line to prevent backflows to protect your fluid system. Enlarging check valves provide a smooth transition from a smaller tubing size to a larger tubing size. Polypropylene body and silicone diaphragm construction offers high purity and exceptional chemical resistance.
The polypropylene body meets USP Plastic Class VI and FDA FDA 21 CFR 177.1520(c) 1.1a requirements, making these check valves ideal for pharmaceutical, food, and beverage applications. The silicone diaphragm opens at 0.5 psi and closes at 1.9 psi ensuring unidirectional flow. A free-floating, disk-based silicone diaphragm allows installation in any position. Single-barb design accepts flexible tubing.
1,400.00 - ₹1,800.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port
Availability Pricing
COMPARE
1/8" hose barb 1/4" hose barb
₹1,600.00INR / Pkg of 10
COMPARE
1/8" hose barb 3/16" hose barb
₹1,500.00INR / Pkg of 10
COMPARE
1/8" hose barb 3/8" hose barb
₹1,400.00INR / Pkg of 10
COMPARE
3/16" hose barb 1/4" hose barb
₹1,800.00INR / Pkg of 10
COMPARE
3/16" hose barb 3/8" hose barb
₹1,500.00INR / Pkg of 10
COMPARE
1/4" hose barb 3/8" hose barb
₹1,700.00INR / Pkg of 10