Cole-Parmer® Enlarging Check Valves, Gamma-Stable Polycarbonate
Safeguard your fluid system from backflows while transitioning diameters
  • Gamma-stable polycarbonate body with silicone diaphragm
  • Free-floating, disk diaphragm allows installation in any position
  • 0.5 psi breaking pressure
  • 1.9 psi sealing pressure
  • Works with both liquids and gases
  • ISO 10993 compliant for biocompatibility
  • DEHP free and RoHS compliant
Place these check valves in your line to prevent backflows to protect your fluid system. Enlarging check valves provide a smooth transition from a smaller tubing size to a larger tubing size. Gamma-stable polycarbonate body and silicone diaphragm construction offers excellent purity and compatibility with gamma irradiation.
The gamma-stable polycarbonate body is ISO 10993 tested for biocompatibility, making these check valves ideal for pharmaceutical and analytical applications. The silicone diaphragm opens at 0.5 psi and closes at 1.9 psi ensuring unidirectional flow. A free-floating, disk-based silicone diaphragm allows installation in any position. Single-barb design accepts flexible tubing.
1,400.00 - ₹1,900.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port
Availability Pricing
COMPARE
1/8" hose barb 1/4" hose barb
₹1,600.00INR / Pkg of 10
COMPARE
1/8" hose barb 3/16" hose barb
₹1,400.00INR / Pkg of 10
COMPARE
1/8" hose barb 3/8" hose barb
₹1,900.00INR / Pkg of 10
COMPARE
3/16" hose barb 1/4" hose barb
₹1,500.00INR / Pkg of 10
COMPARE
3/16" hose barb 3/8" hose barb
₹1,900.00INR / Pkg of 10
COMPARE
1/4" hose barb 3/8" hose barb
₹1,800.00INR / Pkg of 10