Cole-Parmer® EH-200 Series Micro-Kjeldahl Extraction Heater
Specifically designed to determine protein content of food on a micro scale
  • Six-position heater allows for single or multiple digestions
  • Rugged stainless steel construction is durable and easy to clean
  • Dedicated controller for each mantle is safely housed in a "cool zone"
Designed for Kjeldahl digestion on a micro scale, the extraction heaters accommodate up to six 18 to 50 mL round-bottom flasks. Heater includes two support arms that can be mounted on the sides to hold the optional fume collection tube; the tube lets you safely collect and exhaust fumes when not operating the heater under a fume hood.
Each heating mantle has its own independent heat controller, on/off switch, and amber neon indicator light, and can operate up to a maximum element temperature of 800°C. Heat controllers located in the lower part of the unit is safely separated from the upper heating elements by a stainless steel tray and well-ventilated air space. Extra safety features include an earth screen and double fuses on the supply line.
680,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flask Size (mL) Power (VAC) Description
Availability Pricing
COMPARE
50 230 Micro-Kjeldahl Extraction Heater, 600 W, EU Plug; 230 VAC
₹680,000.00INR / Each