Cole-Parmer® Disposable Syringes
Ideal for all syringe filter applications
  • Constructed of durable polypropylene and polyethylene
These all-plastic disposable syringes easily dispense small volumes of liquids. Polypropylene barrel and polyethylene plunger eliminate the need for rubber or synthetic gaskets. No silicone or oil lubricant is required in the syringe barrel. Choose either luer-slip or luer-lock connection type.
14,500.00 - ₹69,900.00INR / Pkg of 100
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Disposable Syringe, Low Deadspace, Luer Slip, 1 mL; 100/Pk
₹31,200.00INR / Pkg of 100
COMPARE
Disposable Syringe, Centric Tip, Luer Slip, 3 mL; 100/Pk
₹14,500.00INR / Pkg of 100
COMPARE
Disposable Syringe, Centric Tip, Luer Slip, 5 mL; 100/Pk
₹38,600.00INR / Pkg of 100
COMPARE
Disposable Syringe, Centric Tip, Luer Slip, 10 mL; 100/Pk
₹25,700.00INR / Pkg of 100
COMPARE
Disposable Syringe, Centric Tip, Luer Slip, 20 mL; 100/Pk
₹35,700.00INR / Pkg of 100
COMPARE
Disposable Syringe, Luer Lock, 5 mL; 100/Pk
₹15,300.00INR / Pkg of 100
COMPARE
Disposable Syringe, Luer Lock, 10 mL; 100/Pk
₹54,600.00INR / Pkg of 100
COMPARE
Disposable Syringe, Luer Lock, 20 mL; 100/Pk
₹69,900.00INR / Pkg of 100
Showing 1- 8 of 8