Cole-Parmer® Disposable 3-Way Stopcocks with Luer Fittings, Polycarbonate
Simply turn the handle to direct the flow
  • Consistent inside diameter minimizes turbulence and bubble entrapment
  • Handle points to "off" port
  • Use with both liquids and gases
  • DEHP free
  • Polycarbonate (PC) housing and polyethylene rotor
Stopcocks provide an easy on/off control of your fluid. Three-way stopcocks feature two female Luer fittings and a male Luer fitting with or without a lock. Luer fittings on stopcocks ensure a quick, leak-free connection. Simply turn the handle to a port you want closed; the other two ports remain open. Stopcocks are made of polyethylene rotor and transparent polycarbonate body, making it easy to see the flow.
1,600.00 - ₹12,700.00INR / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Connection Type Luer Connection Type (Port 3) Quantity
Availability Pricing
COMPARE
Female Luer x Female Luer x Male Locking Luer Male Locking Luer 10
₹1,900.00INR / Pkg of 10
COMPARE
Female Luer x Female Luer x Male Slip Luer Male Slip Luer 10
₹1,600.00INR / Pkg of 10
COMPARE
Female Luer x Female Luer x Male Locking Luer Male Locking Luer 100
₹12,700.00INR / Pkg of 100
COMPARE
Female Luer x Female Luer x Male Slip Luer Male Slip Luer 100
₹11,900.00INR / Pkg of 100