Cole-Parmer® Dewar and Traceable® Temperature Data Logger Bundles
Confidently store and monitor your temperature-sensitive samples
  • Improve sample protection – maintain specific temperature ranges for extended time periods
  • Maintain sample integrity with Traceable® temperature monitoring
  • Data logger provided with 2-year NIST-traceable calibration certificate
The Cole-Parmer® dewar together with a Traceable® data logger is the perfect combination to safely store your critical samples while maintaining sample integrity. Dewars feature durable aluminum construction to hold liquid nitrogen for up to 130 days without refill (depending on model). Included stainless steel racks allow you to store a variety of samples. The bundle adds on one Traceable® Wi-Fi data logger compatible with TraceableLIVE® cloud service (model 18001-08) designed specifically for temperatures down to –200°C. Once your data logger is set up, you can securely monitor parameters 24/7, control up to 100 alarms remotely, view your data logging history, run reports in real time, and give access to other team members.
Enhance sample protection and ease of transportation with these Cole-Parmer® Dewar Accessories which includes Smart Caps, trolleys, and level rulers.
139,000.00 - ₹778,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Static Holding Time (days) Volume (Liters) Neck Inside Diameter (ID) (in)
Availability Pricing
COMPARE
66 95 8 1/2
₹778,000.00INR / Each
COMPARE
71 30 4 7/8
₹249,000.00INR / Each
COMPARE
76 35 4 7/8
₹269,000.00INR / Each
COMPARE
130 47 5
₹321,000.00INR / Each
COMPARE
21 2 1/8
₹139,000.00INR / Each