Cole-Parmer® Desiccators, PP/PC
Ideal working depth for your drying application
  • Rigid, perforated polypropylene plate supports the contents and leaves space for desiccant
Desiccator has a chemical-resistant polypropylene body and polycarbonate cover. A neoprene O-ring ffers a secure seal. Use grease on the flanges to make the unit completely airtight.
25,700.00 - ₹38,500.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Clearance Above Plate (mm) Flange Outside Diameter (OD) (cm) Plate Size (mm)
Availability Pricing
COMPARE
123 17.1 138
₹25,700.00INR / Each
COMPARE
166 23.2 190
₹29,700.00INR / Each
COMPARE
207 27.6 248
₹38,500.00INR / Each