Cole-Parmer® DEHP-Free, Gamma-Stable PVC Tubing
Specifically designed for therapeutic applications such as cell culture and gene therapy
  • Compatible with aseptic welding and sealing machines
  • DEHP, phthalates, and BPA free, and animal-derived ingredient free (ADIF)
  • Crystal clear tubing for easy process monitoring
  • Sterilize by gamma irradiation (<45 kGy), EtO, and E-beam
  • USP Class VI, ISO 10993 Sections 4 and 5), REACH, and RoHS compliant
Specifically designed for cell and gene therapy processes, Cole-Parmer DEHP-free PVC tubing provides unmatched reliability and quality. Superior toxicological and physiochemical profile, gamma stability up to 45 kGy, and compatibility with aseptic welders or sealers, make the tubing perfect for pharmaceutical and bioprocessing environments.
5,600.00 - ₹20,800.00INR / Pkg of 1
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in) Tubing OD (in) Wall Thickness (in)
Availability Pricing
COMPARE
1/16 1/8 1/32
₹8,100.00INR / Pkg of 1
COMPARE
1/8 1/4 1/16
₹11,400.00INR / Pkg of 1
COMPARE
3/16 5/16 1/16
₹5,600.00INR / Pkg of 1
COMPARE
3/16 7/16 1/8
₹10,200.00INR / Pkg of 1
COMPARE
1/4 3/8 1/16
₹6,100.00INR / Pkg of 1
COMPARE
1/4 7/16 3/32
₹9,800.00INR / Pkg of 1
COMPARE
1/4 1/2 1/8
₹11,400.00INR / Pkg of 1
COMPARE
5/16 7/16 1/16
₹10,800.00INR / Pkg of 1
COMPARE
3/8 9/16 3/32
₹13,200.00INR / Pkg of 1
COMPARE
3/8 5/8 1/8
₹15,600.00INR / Pkg of 1
COMPARE
1/2 3/4 1/8
₹20,800.00INR / Pkg of 1
Showing 1- 11 of 11